ContactUsLinkComponent
HPD™ Portable Dispenser, EFD
HPD™ Portable Dispenser, EFD
Catalog # 76243-812
HPD™ Portable Dispenser, EFD
Catalog # 76243-812
Supplier Number:  7023596
Specifications
  • Description:
    HPD™ Portable Dispenser, HPD10K
  • Cat. No.:
    76243-812
  • For use with:
    10 cc Syringe Barrels
Specifications
About this item
This portable fluid dispenser is easy to use and offer durable, fail-safe construction with less hand fatigue.
  • Ergonomic design
  • Features positive shutoff
  • Simplified maintenance
Ideal for touchups, low-volume assembly and field work, our handheld fluid dispensers are compatible with all syringe barrels, pistons and dispensing tips.