Order Entry
United States
ContactUsLinkComponent
Streptavidin, IRDye® 800CW, LI-COR®
Streptavidin, IRDye® 800CW, LI-COR®
Catalog # 102673-342
Supplier:  Li-Cor
CAS Number:  
Streptavidin, IRDye® 800CW, LI-COR®
Catalog # 102673-342
Supplier:  Li-Cor
Supplier Number:  926-32230
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    IRDye® 800CW
  • Size:
    0.5 mg
  • Storage Conditions:
    4 °C
  • Protein/Peptide Name:
    Streptavidin
  • Concentration:
    1 mg/ml
  • Cat. No.:
    102673-342
  • Supplier no.:
    926-32230

Specifications

About this item

IRDye® 800CW Streptavidin can be used as a secondary detection reagent for microscopy, protein arrays, In-Gel westerns, ELISA and western blotting.

  • Dye/protein ratio: 1.8 moles of IRDye® 800CW streptavidin
  • Buffer: 10 mM phosphate, 183 mM NaCl and 2.7 mM KCl, pH 7.4
  • Preservative: Sodium azide (0.005%)

Centrifuge briefly before use to eliminate aggregates that may have formed in solution. This will reduce non-specific background staining.