Dual-Wire Thermoplastic Wrist Strap, SCS, Desco Industries
Catalog # 76238-086
Supplier: Desco
Specifications
- Color:Blue
- Description:Dual Wire Adjustable Thermoplastic Wrist Strap
- Cat. No.:76238-086
Specifications
About this item
The Dual-Wire Thermoplastic Wrist Strap consists of an insulative exterior with a conductive interior and a 3.4 mm dual conductor jack. The wrist strap is lightweight, low profile, and safely makes contact with human skin.
- Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
- Zipper style latching mechanism easily adjusts to most wrist sizes
- Made in the United States of America
Made to be used in conjunction with all SCS Workstation Monitors.
Certifications: Meets requirements of ANSI/ESD S20.20 and Wrist Strap standard ANSI/ESD S1.1.
Ordering information: Designed for use with SCS Dual-Wire Coil Cords (sold separately).