ESD Connector Caps, MSC Series, Caplugs
Catalog # 76253-400
Supplier: Caplugs
Specifications
- Color:Black
- Description:ESD Connector Caps, Shell Size 19
- Cat. No.:76253-400
Specifications
About this item
MSC Series ESD connector caps are manufactured of an ICP (inherently conductive polymer) compound. This non-sloughing material will not mark or crayon when rubbed briskly against a hard, flat, laminated surface.
These non-sloughing caps and plugs, commonly used in military and aerospace, meet the requirements of M 5501/31.