Human Recombinant VLDL R (from HEK293 Cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein Function:Bio-Markers & CD Antigens
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human VLDL R, His Tag (Cat. No. VLR-H5227) at 5 μg/mL (100 μL/well) can bind Biotinylated Human PCSK9, His Tag (Cat. No. PC9-H82E7) with a linear range of 10-156 ng/mL (QC tested).
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:RP11-320E16.1|VLDL receptor|VLDLRCH|FLJ35024|VLDLR|CHRMQ1
- UniProtKB:P98155-2
- Protein/Peptide Name:VLDL R
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:84.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together