Human Recombinant PCSK9 (from HEK293 Cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Bio-Markers & CD Antigens
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 10 mM HCl, pH2.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human LDL R, His Tag (Cat. No. LDR-H5224) at 10 μg/mL (100 μL/well) can bind Human PCSK9, His Tag (Cat. No. PC9-H5226) with a linear range of 39-156 ng/mL (QC tested).
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:HCHOLA3|PCSK9|LDLCQ1|PC9|NARC1|FH3
- UniProtKB:Q8NBP7
- Protein/Peptide Name:PCSK9
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:58 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 10 mM HCl, pH2.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together