Rhesus macaque Recombinant OX40 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Immune Checkpoint Proteins
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Rhesus macaque OX40, His Tag (Cat. No. OX0-C5220) at 2 μg/mL (100 μl/well) can bind Human OX40 Ligand, Fc Tag (Cat. No. OXL-H526x) with a linear range of 0.2-25 ng/mL (QC tested).
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TXGP1L|ACT35|OX40|TNFRSF4|OX40L receptor|CD134
- UniProtKB:XP_001090870.1
- Protein/Peptide Name:OX40
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:22.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together