Human Recombinant IL-17A (from E. coli)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human IL-17A, Tag Free (Cat. No. ILA-H5118) at 2 μg/mL (100 μL/well) can bind Anti-IL17A MAb, Human IgG1 with a linear range of 0.08-1 ng/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Human IL-17A, Tag Free (Cat. No. ILA-H5118) at 0.5 μg/mL (100 μL/well) can bind Human IL-17 RA, His Tag (Cat. No. ILA-H5222) with a linear range of 8-125 ng/mL (QC tested).; Human IL17/ IL17A Protein, Free Tag(Cat. No. ILA-H5118) stimulates secretion of IL-6 by NIH-3T3 cells. The ED50 for this effect is 1.444-6.941 ng/ml.
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IL-17|IL-17A|CTLA-8|Interleukin-17A
- UniProtKB:Q16552
- Protein/Peptide Name:IL-17A
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:15.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT
Frequently Bought Together