Human Recombinant Her2 (from HEK293 cells)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein Function:Bio-Markers & CD Antigens
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human Her2, His Tag (Cat. No. HE2-H5225) at 0.05 μg/mL (100 μL/well) can bind Trastuzumab with a linear range of 0.2-3 ng/mL (QC tested).; Herceptin (Trastuzumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human Her2, His Tag (Cat. No. HE2-H5225) with an affinity constant of 1.07 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Immobilized Herceptin on AHC Biosensor, can bind Human Her2, His Tag (Cat. No. HE2-H5225) with an affinity constant of 0.93 nM as determined in BLI assay (Fortebio Octet 96) (Routinely tested).
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:NEU|MLN19|TKR1|ERBB2|CD340|HER2|HER-2/neu|NGL
- UniProtKB:P04626
- Protein/Peptide Name:Her2
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:70.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT
Frequently Bought Together