Order Entry
United States
ContactUsLinkComponent
HIV Recombinant GP120 (HIV) (from HEK293 cells)
HIV Recombinant GP120 (HIV) (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
HIV Recombinant GP120 (HIV) (from HEK293 cells)
Supplier:  ACRO Biosystems

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    HIV
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 5 μg/mL (100 μL/well) can bind HIV-1 [HIV-1 / Clade E (CM244)] GP120 (Cat. No. GP4-V15223) with a linear range of 20-156 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human CD4, Fc Tag (Cat. No. CD4-H82F3) at 1 μg/mL (100 μL/well) can bind HIV-1 [HIV-1 / Clade E (CM244)] GP120 (Cat. No. GP4-V15223) with a linear range of 0.156-0.313 μg/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human CD4, Fc Tag (Cat. No. CD4-H82F3) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.2ug/well) plate, can bind HIV-1 [HIV-1 / Clade E (CM244)] GP120 (Cat. No. GP4-V15223) with a linear range of 10-78 ng/mL (Routinely tested).
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein/Peptide Name:
    GP120 (HIV)
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    54.1 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together