Avitag™ Cynomolgus macaque;Rhesus macaque Recombinant FcRn (FCGRT & B2M) (from HEK293 cells), Biotin, Acro Biosystems
Supplier: ACRO Biosystems
Certificates
About this item
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkeyRhesus monkey
- Tag sequence:Avi Tag (Avitag™)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Ipilumimab at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus FcRn, His Tag, Strep Tag (Cat. No. FCM-C82W5) with a linear range of 0.02-0.313 μg/mL (QC tested).
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:FcRn|FCGRT & B2M
- UniProtKB:Q8SPV9 (FcRN) & Q8SPW0 (B2M)
- Protein/Peptide Name:FcRn (FCGRT & B2M)
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:34.1 kDa (FCGRT), 13.1 kDa (B2M)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together