Order Entry
United States
ContactUsLinkComponent
Human Recombinant DPP4 (from HEK293 Cells)
Human Recombinant DPP4 (from HEK293 Cells)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant DPP4 (from HEK293 Cells)
Supplier:  ACRO Biosystems

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Enzyme type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Form:
    Powder
  • Shipping Conditions:
    RT
  • Enzyme Name:
    DPP4
  • Protease-free:
    Y
  • Animal-Free:
    Yes
  • Enzyme Synonyms:
    ADABP, ADCP2, CD26, DPPIV, TP103
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    85.4 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM HEPES, 150 mM NaCl, pH 7.5. Normally trehalose is added as protectant before lyophilization.

Specifications

Frequently Bought Together