Human Recombinant E-Selectin (from HEK293 cells)
About this item
2 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 103014-212
- 103014-210
- Protein/Peptide Name:
- E-Selectin
- E-Selectin
- Protein Synonyms:
- E-Selectin|RP1-117P20.2|ELAM1|ESEL|LECAM2|CD62E|SELE
- E-Selectin|RP1-117P20.2|ELAM1|ESEL|LECAM2|CD62E|SELE
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Protein Function:
- Bio-Markers & CD Antigens
- Bio-Markers & CD Antigens
- Species:
- Human
- Human
- Source:
- HEK293 cells
- HEK293 cells
- Conjugation:
- Unconjugated
- Unconjugated
- Protease-free:
- Y
- Y
- Animal-Free:
- Y
- Y
- Endotoxin Level:
- Low
- Low
- Endotoxin Content:
- Less than 1.0 EU per μg by the LAL method.
- Less than 1.0 EU per μg by the LAL method.
- UniProtKB:
- P16581
- P16581
- Biological Activity:
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Purity:
- >95% as determined by SDS-PAGE.
- >95% as determined by SDS-PAGE.
- Formulation:
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Molecular Weight:
- 60 kDa
- 60 kDa
- Storage Conditions:
- –20 °C
- –20 °C
- Shipping Temperature:
- Powder,RT
- Powder,RT
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: CDEH5222200UGSpecifications:Marketsource
More
Specifications
Cat. No.103014-212Protein/Peptide NameE-SelectinProtein SynonymsE-Selectin|RP1-117P20.2|ELAM1|ESEL|LECAM2|CD62E|SELEProtein/Peptide TypeRecombinantProtein FunctionBio-Markers & CD AntigensSpeciesHumanSourceHEK293 cellsConjugationUnconjugatedProtease-freeYAnimal-FreeYEndotoxin LevelLowEndotoxin ContentLess than 1.0 EU per μg by the LAL method.UniProtKBP16581Biological ActivityLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.Purity>95% as determined by SDS-PAGE.FormulationLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.Molecular Weight60 kDaStorage Conditions–20 °CShipping TemperaturePowder,RTMarketsource - Supplier Number: CDEH52221MGSpecifications:Marketsource
More
Specifications
Cat. No.103014-210Protein/Peptide NameE-SelectinProtein SynonymsE-Selectin|RP1-117P20.2|ELAM1|ESEL|LECAM2|CD62E|SELEProtein/Peptide TypeRecombinantProtein FunctionBio-Markers & CD AntigensSpeciesHumanSourceHEK293 cellsConjugationUnconjugatedProtease-freeYAnimal-FreeYEndotoxin LevelLowEndotoxin ContentLess than 1.0 EU per μg by the LAL method.UniProtKBP16581Biological ActivityLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.Purity>95% as determined by SDS-PAGE.FormulationLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.Molecular Weight60 kDaStorage Conditions–20 °CShipping TemperaturePowder,RTMarketsource
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



