Human Recombinant CD19 (from HEK293 cells), Biotin, Fc Tag
Supplier: ACRO Biosystems
Certificates
About this item
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-Mouse FMC63 Mab (Cat. No. CD9-M7) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD19, Fc Tag (Cat. No. CD9-H8259) with a linear range of 0.039-0.625 μg/mL (QC tested).; 293 cells were transfected with FCM63-scFv and RFP tag . 2x105 of the cells were first incubated with A. Biotinylated protein control. B. Recombinant biotinylated human CD19 (Cat. No. CD9-H8259, 10 μg/ml ) . C. Recombinant biotinylated human CD19 (Cat. No. CD9-H8259, 10 μg/ml) and FMC63(Mouse anti-CD19 antibody). FITC Streptavidin was used to analyse with FACS. RFP was used to evaluate CAR(FMC63-scFv) expression and FITC was used to evaluate the binding activity of recombinant biotinylated human CD19 (Cat. No. CD9-H8259) .; Human T cells were lentivirally transduced with anti-CD19 CAR and cultured for 11 days. Eleven days post-transduction, 1e6 cells were stained for the expression of CD3 and anti-CD19 CAR with FITC anti-human CD3 antibody and biotinylated human CD19 (Cat. No. CD9-H8259) followed by PE-conjugated streptavidin, respectively. A. Non-transduced T cells were used as a control for gating of CAR expression. (Data are kindly provided by Beijing Bowei Huaen Medical Technology Co. Ltd.)
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CVID3|MGC12802|B4|CD19
- UniProtKB:P15391
- Protein/Peptide Name:CD19
- Purity:>95% as determined by reduced SDS-PAGE.
- Molecular Weight:56.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together