Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant B7-1 (from HEK293 cells), Fc Tag
Mouse Recombinant B7-1 (from HEK293 cells), Fc Tag
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse Recombinant B7-1 (from HEK293 cells), Fc Tag
Supplier:  ACROBIOSYSTEMS

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immune Checkpoint Proteins
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    Fc Tag
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse B7-1, Fc Tag (Cat. No. CD0-M5259) at 2 μg/mL (100 µl/well),can bind Mouse CTLA-4, His Tag (Cat. No. CT4-M52H5) with a linear range of 0.02-0.8 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Mouse B7-1, Fc Tag (Cat. No. CD0-M5259) at 5 μg/mL (100 μL/well) can bind Mouse PD-L1, mouse IgG2a Fc tag, low endotoxin (Cat. No. PD1-M52A2) with a linear range of 0.16-2.5 μg/mL (Routinely tested).
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    B7-1|B7.1|CD28LG|CD28LG1|LAB7|BB1|CD80|B7
  • UniProtKB:
    Q00609-1
  • Protein/Peptide Name:
    B7-1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    50.3 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together