Order Entry
United States
ContactUsLinkComponent
Human Recombinant Carbonic Anhydrase II (from HEK293 cells)
Human Recombinant Carbonic Anhydrase II (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant Carbonic Anhydrase II (from HEK293 cells)
Supplier:  ACRO Biosystems

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Bio-Markers & CD Antigens
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 20 mM Tris, pH 8.0 with 150 mM NaCl, 1 mM DTT. Normally trehalose is added as protectant before lyophilization.
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CA2|CA-II|Car2|CAII
  • UniProtKB:
    P00918
  • Protein/Peptide Name:
    Carbonic Anhydrase II
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    30 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 20 mM Tris, pH 8.0 with 150 mM NaCl, 1 mM DTT. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together