Human Recombinant BACE1 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Fc Tag fused with human IgG1 Fc tag at C-terminus
- UniProtKB: P56817
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Enzyme type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage Conditions:−20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Form:Powder
- Shipping Conditions:RT
- Enzyme Name:BACE1
- Protease-free:Y
- Animal-Free:Yes
- Enzyme Synonyms:ASP2, BACE, FLJ90568, HSPC104, KIAA1149, Memapsin-2
- Purity:>85% as determined by SDS-PAGE.
- Molecular Weight:75 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7.5. Normally trehalose is added as protectant before lyophilization.
Specifications
Frequently Bought Together