ContactUsLinkComponent
Desiccant in Tyvek® Pouch, SCS Division of Desco
Desiccant in Tyvek® Pouch, SCS Division of Desco
Catalog # 76247-632
Supplier:  Desco
Desiccant in Tyvek® Pouch, SCS Division of Desco
Catalog # 76247-632
Supplier:  Desco
Supplier Number:  2PLDES150
Specifications
  • Description:
    Desiccant, 2 units
  • Size:
    150/pack
  • Environmentally Preferable:
  • Cat. No.:
    76247-632
  • Packaging:
    Tyvek® Pouch
Specifications
About this item
These dessicants provide protection for moisture sensitive electronics (such as SMT – Surface Mount Technology).
  • Secured in strong, sulphur-free non-woven plastic pouches
  • Can be baked and re-used
  • No hazardous substances
  • Recommended for use in clean rooms
Recommended amount of desiccant is dependent on the interior surface area of the bag to be used.