Semco® Legacy Red Scraper, Bergdahl Associates
Catalog # 76240-836
Supplier: BERGDAHL ASSOCIATES INC. TE
Specifications
- Description:Legacy Red Scraper
- Cat. No.:76240-836
Specifications
About this item
This scraper is fabricated from Celcon® POM and will not scratch or score aluminum substrates.
Certifications: The tool has been evaluated and approved by Boeing for the removal of cured sealant and is listed in Boeing BAC 5000 process specification.