Loctite® Adhesives, R.S. Hughes, Henkel
Supplier: RS HUGHES TE
Certificates
About this item
These Loctite clear cyanoacrylate adhesives are compatible with metal, thermoplastic and thermoset plastic materials with a 24 hour cure time.
- Delivers great performance
Packaging: Packed in a 20 g bottle, 10 per case.
Specifications
- Catalog No:
- 76252-544
- 76252-546
- Temperature Range:
- –65...+160 °F
- –65...+220 °F
- Tensile Strength:
- 2030 - 3190 psi
- 2755 psi
Specifications
Frequently Bought Together