ContactUsLinkComponent
Laminate Grounding System, SCS Division of Desco
Laminate Grounding System, SCS Division of Desco
Catalog # 76241-462
Supplier:  Desco
Laminate Grounding System, SCS Division of Desco
Catalog # 76241-462
Supplier:  Desco
Supplier Number:  3044
Specifications
  • Description:
    Laminate Grounding System
  • Cat. No.:
    76241-462
Specifications
About this item
This is a common point ground system for benches with laminate tops.
Delivery: Includes one 10' (3.0 m) grounding wire with one megohm resistor and ring terminal, and one grounding bolt assembly with single wrist strap banana jack connection terminal.