Bottle and Can Recycling Top, Global Industrial
Catalog # 76249-460
Supplier: GLOBAL INDUSTRIAL TE
Specifications
- Depth:11⁵⁄₁₆"
- Width:20⅜"
- Color:Brown
- Description:Bottle and Can Recycling Top
- Height:2¾"
- Environmentally Preferable:
- Cat. No.:76249-460
- For use with:Slim Jim Recycling Containers
Specifications
About this item
Bottle and can recycling top for rubbermaid slim jim recycling containers.
- Durable plastic construction is long lasting
- Lid fits both the 16 and 23 gallon containers (sold separately)
- Eliminates the need for messy sorting
This lid features 2 round openings to accommodate recyclable bottles and cans while keeping out non recyclable waste.