Aurelia® Ignite™ Nitrile Gloves, Supermax Healthcare
Supplier: Avantor
Certificates
About this item
Aurelia Ignite™ orange nitrile gloves feature our patented MAX Grip™ no slip texture.
- High visibility orange color
- Excellent puncture resistance
- Superior chemical resistance and tensile strength
- Tested and approved for Fentanyl resistance per ASTM D6978-05 standards
Frequently Bought Together