ContactUsLinkComponent
Weller® Precision 125 mm Tweezer Pointed Tip, Apex Tools
Weller® Precision 125 mm Tweezer Pointed Tip, Apex Tools
Catalog # 76238-628
Supplier:  Cooper Tools
Weller® Precision 125 mm Tweezer Pointed Tip, Apex Tools
Catalog # 76238-628
Supplier:  Cooper Tools
Supplier Number:  AM
Specifications
  • Length:
    125 mm
  • Material:
    Brass
  • Cat. No.:
    76238-628
  • Tip style:
    Pointed
Specifications
About this item
Product made from brass.
  • Suitable for delicate standard applications and precision work on small components or wires
  • The soft metal protects sensitive components against damage
  • No sparks
For applications in microelectronics, jewelry making, watch making, medicine, and laboratory technology.