Weller® Precision Tweezer with Fine But Robust Tips, Apex Tool Group
76238-626
- Length:125 mm
- Material:Special Stainless Steel
- Cat. No.:76238-626
- Tip style:Robust
Designed for standard applications.
- Suitable for delicate standard applications
- Precision work on small components or wires
The special stainless steel is non-magnetic. The tweezers are non-rusting, acid-proof and heat resistant up to 300 °C (512 °F). Tweezers made from special stainless steel have the identification letter "SA" in their order numbers. For applications in microelectronics, jewelry making, watch making, medicine, and laboratory technology.