Orders LinkContactUsLinkComponent
Embossed Copper Foil Shielding Tapes, 3M
Embossed Copper Foil Shielding Tapes, 3M
Supplier:   3M
Embossed Copper Foil Shielding Tapes, 3M
Supplier:  3M
Certificates
Embossed Copper Foil Shielding Tapes, 3M
2 Options Available Below
Product Details & Documents
In these types of tapes 4 mil copper foil backing with acrylic pressure-sensitive adhesive and liner for good shielding effectiveness, soldering and converting.
  • Embossing makes excellent contact through the adhesive
  • The un-plated copper is for non-corrosive conditions
  • Removable liner makes it easy to handle and die cut complex shapes
Embossed pattern through adhesive provides very low contact resistance between the substrate and the backing to drain static charge.
Certifications: UL recognized flame-retardancy per UL 510 for applications needing this property.
Product Family Options
Product Information
  • Length
    Width
    Thickness
    Packaging
    Availability
    Price
  • 76242-672
    Embossed Copper Foil Shielding Tapes
    Embossed Copper Foil Shielding Tapes
    54'
    ⅜"
    4 mil.
    24 Rolls/Case
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $42.40
    $42.40
    Log in for availability
    Case of 24
    Case of 24 - $1,017.60
    $1,017.60
    Each
       $42.40
    Case of 24
       $1,017.60
    Supplier Number: 7010350291
  • 76244-818
    Embossed Copper Foil Shielding Tapes
    Embossed Copper Foil Shielding Tapes
    54'
    2"
    4 mil.
    5 Rolls/Case
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $238.00
    $238.00
    Log in for availability
    Case of 5
    Case of 5 - $1,190.00
    $1,190.00
    Each
       $238.00
    Case of 5
       $1,190.00
    Supplier Number: 7010304004