ContactUsLinkComponent
Chucking Reamer, Cutting Tool Supply
Chucking Reamer, Cutting Tool Supply
  76253-664
Chucking Reamer, Cutting Tool Supply
  76253-664
 :  00084-AP
  • Description:
    Chucking Reamer
  • Length:
    4½"
  • Size:
    3/16"
  • Cat. No.:
    76253-664
  • Shank diameter:
    0.1805 - 0.1795"
  • Flute length:
    1⅛"
Made of high speed steel.
  • With straight shank and flute