Grounding Cord for ESD Floor Mats, Ranco Industries
Catalog # 76253-702
Supplier: RANCO INDUSTRIES INC. TE
Specifications
- Description:Grounding Cord
- Cat. No.:76253-702
Specifications
About this item
Grounding cord to connect all ESD mats to a building ground or an electrical outlet, using the screw that secures the outlet cover as the attachment point.