Kelvin Test Probe Set, Newark/Element
Catalog # 76249-630
Supplier: NEWARK/ELEMENT 14 TE
Specifications
- Description:Test Lead, BNC Connector to Kelvin Probe Set
- Environmentally Preferable:
- Cat. No.:76249-630
Specifications
About this item
Kelvin Test Probe Set consists of RG-174 cable with a BNC connector on one end and a Kelvin probe on the other end. Spring contacts of the Kelvin probe are sharply pointed to penetrate corrosion and are close together, for usage in tight areas.