Order Entry
United States
ContactUsLinkComponent
 
Bovine Collagen Type I (Methacrylated)
Catalog # 103701-730
CAS Number:  
undefined
Bovine Collagen Type I (Methacrylated)
Catalog # 103701-730
Supplier Number:  5198-100MG
CAS Number:  

Specifications

  • Species:
    Bovine
  • Size:
    100 mg
  • Protein/Peptide Name:
    Collagen Type I (methacrylated)
  • Formulation:
    Lyophilized
  • Cat. No.:
    103701-730

Specifications

About this item

Advanced BioMatrix offers PhotoCol®, a purified methacrylated Type I bovine collagen kit. PhotoCol® provides native-like 3D collagen gels with the unique attributes to be prepared at various concentrations and crosslinked to provide various gel stiffness.

The PhotoCol® kit consists of purified methacrylated Type I bovine collagen as the core component with other support reagents in the kit.

The methacrylated Type I collagen is produced from telo-peptide intact bovine collagen where the collagen has been modified by reacting the free amines, primarily the e-amines groups of the lysine residues as well as the a-amines groups on the N-termini. Approximately 40% of the total lysine residues of the collagen molecule have been methacrylated. The collagen is extracted from bovine hide and contains a high monomer content. The collagen starting material was isolated from a closed herd and purified using controlled manufacturing processes. A bottle of 20 mM acetic acid solution is provided to solubilize the lyophilized methacrylated collagen at concentrations ranging from 3 to 8 mg/ml.The neutralization solution consists of an alkaline 10X phosphate buffered saline (PBS) solution which provides physiological salts and pH in the final mixture.The photoinitiator consists of Irgacure 2959 to be formulated in methanol (methanol not included) which allows UV crosslinking of the collagen at 365 nm.

  • Highly Purified
  • Lyophilized to allow you to choose the desired concentration
  • Photocrosslinkable with a wide variety of photoinitiators
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.