To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Source:
- Species:Bovine
- Size:0.5 mg
- Environmentally Preferable:
- Protein/Peptide Name:Collagen Type II
- Concentration:0.5 mg/mL
- Cat. No.:103701-728
Bovine Collagen Type II is one of a family of proteins found particularly in the flesh and connective tissues of mammals (approximately one-third of the body’s total protein). Over two dozen types of collagen have been described; Type II is found predominantly in hyaline cartilage (50% of the protein in hyaline is collagen type II) and is also found in the vitreous humor of the eye.
The product is supplied as a sterile solution at approximately 0.5 mg per vial at approximately ~.50 mg/ml in 500 mM acetic acid. A certificate of Analysis is available with the purchase of each product. Type II collagen is provided in user-friendly packaging for use and storage. Type II collagen may be used to culture a variety of cell types. Type II collagen is typically used as a coating material for cell culture studies, for the formation of 3D collagen gels, or as an antigen for antibody production.
- Great for 2D Coatings
- Highly purified collagen
- Over 15 QC tests to ensure consistency and quality