Poly-L-Lysine
Catalog # 103701-228
Supplier: Avantor
CAS Number:
Specifications
- Size:5 mg
- Form:Powder
- Protein/Peptide Name:Poly-L-Lysine
- Molecular Weight:70,000 to 150,000 Da
- Cat. No.:103701-228
Specifications
About this item
Poly-L-Lysine is a synthetic amino acid chain that is positively charged and widely used as a coating to enhance cell attachment and adhesion to both plasticware and glass surfaces.
The molecular weight of Poly-L-Lysine can vary significantly with lower molecular weight (30,000 Da) being less viscous and higher molecular weight (>300,000 Da) having more binding sites per molecule. This product’s molecular weight ranges from 70,000 to 150,000 Da.
Poly-L-Lysine is supplied in a 50 ml amber bottle containing 5 mg of Poly-L-Lysine. The product has been sterilized and is ready-to-use after proper addition of cell culture water.
- Enhances cell attachment to plasticware and glass
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.