- Size:50 ml
- Form:Solution
- Protein/Peptide Name:Poly-L-Ornithine
- Molecular Weight:70,000 to 150,000 Da
- Concentration:0.1 mg/ mL
- Cat. No.:103701-204
Poly-L-Ornithine is a synthetic amino acid chain that is positively charged having one hydrobromide per unit of ornithine. Poly-L-Ornithine is widely used as a coating to enhance cell attachment and adhesion to both plasticware and glass surfaces.
The molecular weight of Poly-L-Ornithine can vary significantly with lower molecular weight (30,000 Da) being less viscous and higher molecular weight (>300,000 Da) having more binding sites per molecule. This product’s molecular weight ranges from 70,000 to 150,000 Da yielding a solution viscosity for easy handling while providing sufficient binding sites for cell attachment.
Poly-L-Ornithine surface coatings are designed to improve cell attachment, growth and differentiation of many cell types. Coated surfaces will often improve cell attachment in reduced or serum-free conditions. This product is supplied in a sterile 50 ml package size at a concentration of 0.1 mg/ml.
- Enhances cell attachment to plasticware and glass