- Species:Human
- Size:1 mg
- Protein/Peptide Name:Fibronectin
- Cat. No.:103701-196
Fibronectin is a widely used broad range natural cell attachment factor. This product has been purified from human plasma where it is found as a dimer and has a size of 440-500 kDa with two similar subunits (220-250 kDa ) linked by two disulfide bonds. Fibronectin’s primary function is related to cell adhesion and attachment to extracellular matrix. Certain domains of Fibronectin have been found to play important roles that involve interactions with collagen, heparin and other cell surface glycosaminoglycans (GAGs). Fibronectin is ideal for coating of surfaces and is typically used at a concentration of 1-5 ug/cm2 . It is provided in user-friendly packaging for use and storage. Fibronectin is sterile filtered and is supplied as a ready to use solution after thawing and concentration adjustment. This product is shipped separately on dry ice.
- Great for 2D coatings
- Highly purified, native ECM
- Known for purity, consistency and quality
- Purified from human plasma