Specifications
- Species:Bovine
- Size:15 mg
- Protein/Peptide Name:Collagen
- Formulation:Lyophilized
- Cat. No.:103700-640
Specifications
About this item
PureCol® collagen is known as the standard of all collagens for purity (>99.9% collagen content), functionality, and the most native-like collagen available. PureCol® is isolated from bovine hides sourced from the only controlled, closed herd in the United States. Advanced BioMatrix’s manufacturing processes comply with stringent quality standards that have proven to yield unsurpassed lot-to-lot consistency.
PureCol® collagen is approximately 97% Type I with the remainder being comprised of Type III collagen. It is soluble atelo-collagen. It contains high monomer content as measured by gel permeation chromatography.
This product is supplied as a lyophilized powder with 15 mg of bovine collagen. When reconstituted with 5 ml of sterile 0.01 N HCl, a concentration of approximately 3 mg/ml is achieved.
PureCol® collagen is ideal for coating of surfaces and providing preparations of thin layers for culturing cells. PureCol®, lyophilized form is not recommended for the formation of a solid gel. PureCol® collagen is provided in user-friendly packaging for use and storage. This product is supplied as a sterile lyophilized powder.
- Great for 2D coatings
- Highly purified, native ECM
- Known for purity, consistency and quality