Wire Gauge Size Short-Flute Cobalt Steel Drill Bit, McMaster Carr
76245-022
- Length:2⅜"
- Material:Black Oxide Cobalt Steel
- Size:#40
- Cat. No.:76245-022
- Shank diameter:0.098"
Jobbers' length are the most common bits and have the rigidity and length required for most applications.
- A split point keeps the bit centered without a pilot hole
- Short flutes provide increased rigidity to drill straighter holes
Suitable for steel, iron, nickel, titanium and stainless steel.