Order Entry
United States
ContactUsLinkComponent
Universal Mounting Bar, Production Basics
Universal Mounting Bar, Production Basics
Catalog # 76246-990
Supplier:  Production Basics
Universal Mounting Bar, Production Basics
Catalog # 76246-990
Supplier:  Production Basics
Supplier Number:  PB-8676

Specifications

  • Color:
    Black
  • Cat. No.:
    76246-990
  • Weight:
    10 lbs.
  • Width:
    48"

Specifications

About this item

Use the universal mounting bar to add tablets or large monitors or multiple monitors to your workstation.

  • Made of 11 gauge steel

For use with one or more flat screen monitor brackets or the tablet holder bracket.