ContactUsLinkComponent
XYZ-Axis Electrically Conductive Tape, 3M
XYZ-Axis Electrically Conductive Tape, 3M
  76248-200
 :  3M
XYZ-Axis Electrically Conductive Tape, 3M
  76248-200
 :  3M
 :  7000048870
  • Width:
    1"
  • Description:
    XYZ-Axis Electrically Conductive Tape
  • Length:
    108'
  • Thickness:
    3 mil.
  • Cat. No.:
    76248-200
XYZ-Axis is a double sided, isotropically conductive pressure sensitive tape which conducts electricity through the thickness (the Z-axis) and the plane of the adhesive (X, Y planes).
  • Provides good adhesion and electrical performance
  • Isotropic XYZ-axis electrical connectivity
  • Good contact with both hard and soft surfaces
Excellent die-cutting and converting capabilities and can be applied in strips or die cut to specific shapes and sizes.