ContactUsLinkComponent
Scotch® Vinyl Electrical Tapes 33, 3M™
  76243-022
 :  3M
Scotch® Vinyl Electrical Tapes 33, 3M™
  76243-022
 :  3M
 :  7000057496
  • Width:
    0.5"
  • Color:
    Black
  • Description:
    Scotch® Vinyl Electrical Tapes 33
  • Length:
    108'
  • Thickness:
    7 mil.
  • Cat. No.:
    76243-022
Scotch® Tapes 33 are mid-tier, vinyl electrical tape that provides electrical insulation for all wires and cables rated up to 600 V. These are compatible with solid dielectric cable insulation, synthetic cable insulations, jackets and splicing compounds.
  • Resists UV rays, abrasion, moisture, alkalis, solvents and acids
  • Provides a protective jacket for high-voltage cable splices
The combination of the pressure-sensitive, rubber-resin adhesive and the PVC backing provides electrical and mechanical protection. These are designed for above/below grade, indoor/outdoor, low- and high-voltage applications and for harnessing cables.
 : UL listed, CSA certified and RoHS 2011/65/EU compliant.