Order Entry
United States
ContactUsLinkComponent
Hi-DTuff High Density Can Liners, Sunland Manufacturing
Hi-DTuff High Density Can Liners, Sunland Manufacturing
Catalog # 76253-978
Hi-DTuff High Density Can Liners, Sunland Manufacturing
Catalog # 76253-978
Supplier Number:  40X48

Specifications

  • Capacity:
    40 - 45 gal.
  • Color:
    Natural
  • Description:
    Hi-DTuff high density can liners, 13 micron
  • Cat. No.:
    76253-978
  • W×L:
    40×48"

Specifications

About this item

The Hi-DTuff is all purpose high density liners.

  • No side seams or side seals
  • Leak resistant, star seal bottom
  • Great for odor confinement
  • Source reduction vs. low density can liners
  • Resins are USDA and FDA compliant