iTorque Screwdriver Handle, Wiha Tools
Catalog # 76249-958
Supplier: Avantor
Specifications
- Description:TorquControl Adjustable iTorque Handle 1.0 - 6.0 Nm, 9 - 50 in./lbs.
- Cat. No.:76249-958
Specifications
About this item
TorquControl Adjustable iTorque Handle.
- Handle reads Ncm, inch and ounces
- Built-in click counter
- Auto shut off
Fast adjusting with interchangeable blades or bits and about ±6% accuracy.
Delivery information: Includes 4 mm Bit Holder.