IBC™ Cleaning Tools, MT Series, US Conec LTD
Supplier: US CONEC LTD. CUSTOMER SERVICE TE
Certificates
About this item
The IBC™ Cleaners MT series mechanical cleaning tools are designed for cleaning the fiber arrays on MT based connector systems.
- Cleans male and female MT ferrule
- Simple push style or dial turn engagement
- Alignment cap lid opens for cleaning the unmated connectors
- Cleans male (with guide pins) and female MT ferrules
These tools are capable of cleaning MT based connector systems loaded in a bulkhead adapter, on the card edge, the backplane and on unmated cable assemblies.
It cleans clean both female (no guide pin) and male (with steel guide pins). These tools will clean both flat polished multimode and 8˚ angled singlemode MT ferrules.
Certifications: RoHS compliant.
Frequently Bought Together