IBC™ Cleaning Tools, Standard Series, US Conec LTD
The IBC™ Cleaners are mechanical cleaning tools designed to clean connectors residing in an adapter or faceplate and unmated connectors.
- Crush resistant to over 250 N
- Impact resistant to survive drops over 1.5 M
- Simple pushing motion to engage tool
The IBC™ Brand cleaning tools use a novel dry cleaning strand to gently sweep and lift away dust and residues from the connector end face. Dry cleaning strand eliminates the need for solvents to be effective.
Frequently Bought Together