ContactUsLinkComponent
Pure-Fit® Large Tube Clamps, Holland Applied Technologies
Pure-Fit® Large Tube Clamps, Holland Applied Technologies
Catalog # 76240-126
Pure-Fit® Large Tube Clamps, Holland Applied Technologies
Catalog # 76240-126
Supplier Number:  PFTC-375-PP
Specifications
  • Description:
    Pure-Fit® TC Selection Guide
  • Material:
    PP
  • Cat. No.:
    76240-126
  • Opening dimensions:
    1/8 - 3/8"
Specifications
About this item
Pure-Fit® TCL Large Tube Clamps provide the same innovative clamping technology as Pure-Fit® TC, but are designed to accommodate large diameter and thick wall tubing applications. Pure-Fit® TCL is compatible with standard, braided (reinforced) and multilayer tubing. In addition to providing complete fluid stoppage, Pure-Fit® TCL can be utilized in the flow position and features an integral bore hole for absolute lock-out capabilities.
  • Can be utilized in the flow position
  • Integral bore hole for lock-out capabilities
  • Compatible with large diameter, thick wall, reinforced and multilayer tubing
  • Provides complete fluid stoppage
  • Can be installed on or over tubing or finished assemblies
Pure-Fit® TCL is constructed from animal derivative-free and BPA-free PVDF. Pure-Fit® TCL is available in two sizes to accommodate a wide range of thick wall tubing and can be used with up to 2.0" O.D. tubing.