ContactUsLinkComponent
Flex500™ HVC Series Flexible Round Caps, Caplugs
Flex500™ HVC Series Flexible Round Caps, Caplugs
Catalog # 76245-386
Supplier:  Caplugs
Flex500™ HVC Series Flexible Round Caps, Caplugs
Catalog # 76245-386
Supplier:  Caplugs
Supplier Number:  HVC-3625-18
Specifications
  • Color:
    Black
  • Dimensions:
    3.625×3.785×1.125" (92.08×96.14×28.58 mm)
  • Cat. No.:
    76245-386
Specifications
About this item
The HVC series Flex 500™ vinyl caps are temperature-resistant and protect up to 500 °F. They are extremely flexible and durable protective closures which are ideal for use as thread protectors, dust and moisture seals, and act as masking caps during painting, plating or powder coating operations.