Low-Profile Polyimide Labels for Thermal Transfer Printers, Type B-729, Brady
Supplier: Brady Worldwide
Certificates
About this item
Low-Profile Polyimide (B-729) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labeling.
- Service temperature of −94 to 212 °F (–70 to 100 °C)
- Ultra-durable matte white polyimide material is thermal transfer printable
- High temperature low profile is ideal for processes requiring thin or lighter weight materials
For use with BBP®81 label printer, BRADY IP® series label printer, BradyPrinter i5100, BradyPrinter i7100, BradyPrinter PR Plus series label printer, THT 3" core printer.
Resistant to abrasion, chemical, corrosion, humidity, UV and weather.
Certifications: RoHS compliant to 2011/65/EU; UL approved; Meets requirements of MIL-STD-202G, Method 215K.
Frequently Bought Together