Phillips® Screwdriver, with SoftFinish® Handle, Wiha Tools
Supplier: WIHA TOOLS TE
Certificates
About this item
These screwdrivers are possessing round blade which protection against electrostatic discharge.
- Made of high alloy CVM steel, hardened, hard chromed
Features with dissipative SoftFinish® cushion grip handle.
Certifications: DIN EN 61340-5-1/61340-5-2 R certified.
Frequently Bought Together