Tape 9490LE, Double Coated, Tapecase
Catalog # 76248-372
Supplier: TAPECASE LTD. TE
Specifications
- Width:½"
- Color:Tan
- Length:60 yds.
- Thickness:6.7 mil.
- Cat. No.:76248-372
Specifications
About this item
Double Coated Tape 9490LE with 3M™ laminating adhesive 300LSE provides high bond strength to most surfaces, including many low surface energy plastics such as polypropylene and powder coated paints.
- For battery bonding, battery door attachment
- Rubber feet bonding
- Touchpad bracket assembly
This tape has a film carrier which can add dimensional stability to foams and other substrates and also make it easier to handle the tape during slitting and die-cutting.