ContactUsLinkComponent
8200 Series 3-Layer Vinyl Floor Mat Roll, Menda
8200 Series 3-Layer Vinyl Floor Mat Roll, Menda
  76251-518
 :  Desco
undefined
8200 Series 3-Layer Vinyl Floor Mat Roll, Menda
  76251-518
 :  Desco
 :  8254
  • Color:
    Blue
  • Length:
    732 cm (24 ft.)
  • Thickness:
    3.2 mm (⅛")
  • Cat. No.:
    76251-518
  • Width:
    122 cm (4 ft.)
Mat made of three layer vinyl material provides reliable path to ground and eliminates static charge generation as a worksurface table mat, shelf liner and cart liner.
  • Highly, conductive scrim middle layer provides main discharge path to ground
  • Dissipative foam bottom layer provides durable, nonskid cushion, choice for components susceptible to physical shock
 : Meets ANSI/ESD S20.20, ANSI/ESD S4.1.