Order Entry
United States
ContactUsLinkComponent
Super-Tack™ Construction Tape, Ultratape Industries
Super-Tack™ Construction Tape, Ultratape Industries
Catalog # 76240-852
Super-Tack™ Construction Tape, Ultratape Industries
Catalog # 76240-852
Supplier Number:  1160WH100-P3D

Specifications

  • Temperature range:
    –20...190 °F
  • Description:
    Super-Tack™ Construction Tape, Rubber Adhesive with Vinyl Backing
  • Length:
    36 yards
  • Thickness:
    6 ±0.7 mil.
  • Cat. No.:
    76240-852
  • Elongation:
    150%
  • Tensile strength:
    18 lbs./in.

Specifications

About this item

UltraTape Construction tapes are designed for critical environment construction and support applications. They can be used for hanging heavy plastic sheeting to create temporary walls, taping cart wheels and ductwork, floor marking, and color coding. Use the colored, striped hazard warning tapes to designate the OSHA critical areas in cleanrooms.

  • Super-Tack™ construction tape has a strong adhesive bond that leaves no residue upon removal
  • Protocol™ tapes for immediate recognition that certified cleanroom compatible tape is in use
Delivery: Available in UltraClean and standard clean.